Learn More
Leica Biosystems 7mL p16 (6H12) BOND RTU Primar
Supplier:  Leica Biosystems PA0016U
Encompass_Preferred
Antigen Backgroundp16 (INK4a) (Cyclin-dependent kinase inhibitor 2A (CDKN2A)) is a tumour suppressor protein associated with cell cycle progression, specifically in the regulation of transition from G1 phase of the cell cycle in to the S phase. Oncogenic mutations in the CDKN2A gene that encodes p16 (resulting in over or under expression of the protein) is associated with a wide range of cancers and cancer precursor lesions.Disclaimerp16 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2183
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.