Learn More
Leica Biosystems 7ML SOX-11 (MRQ-58 Mab) 'CM'
Supplier:  Leica Biosystems PA0816
Encompass
This antibody is intended for in vitro diagnostic (IVD) use. The Cell Marque SOX-11 (MRQ-58) antibody is intended for qualified laboratories to qualitatively identify by light microscopy the presence of associated antigens in sections of formalin-fixed, paraffin-embedded tissue sections using IHC test methods. Use of this antibody is indicated, subsequent to clinical differential diagnosis, as an aid in the identification and differentiation of mantle cell lymphomas within the context of an antibody panel, the patient’s clinical history, and other diagnostic tests evaluated by a qualified pathologist.
Catalog No. 50-255-2030
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.