Learn More
Leica Biosystems 7mL TIM-3 (EPR22241) BOND RTU
Supplier:  Leica Biosystems PA0360U
Encompass
Antigen BackgroundTIM-3 (T-cell Immunoglobulin and Mucin Domain-Containing 3, Hepatitis A Virus Cellular Receptor, HAVCR2). TIM-3 is a Type I transmembrane that can be expressed on both tumour and immune cells. It is expressed on multiple immune cells, including type 1 T helper (Th1) cells, Th17 and CD8+ T cells, Tumour Infiltrating Lymphocytes, regulatory T cells (Tregs) and innate immune cells.DisclaimerTIM-3 is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2105
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.