Learn More
Description
ES 7X-O-Matic is a powerful wetting and penetrating agent, which contains a non-ionic surfactant and a hydro-trope to complex phosphates. It will penetrate and clean minute crevices not accessible to ordinary cleaners and is immediately and completely soluble in water at any concentration, without agitation. ES 7X-O-Matic was designed to clean under pressure, heat and agitation. It behaves and remains mostly suds-free when agitated.
With normal detergents the precipitation of solids on cleaned surfaces is a major problem. The powerful dispersing agents in the 7X solutions work to reduce agglomerates to individual particles and hold them in suspension so that they are not re-deposited on the surfaces being cleaned.
Laboratory equipment and glassware washed in any of the 7X solutions drain completely in a few seconds. Solutions which do not drain quickly can allow molecules of solute to be deposited on the washed surface to the extent that a visible film is present. Such deposits can affect critical experimental work.
7X cleaning solutions are compatible with alkalis and acids. The pH is near normal and varies only minimally with the concentration. The neutral pH levels will not induce etching of delicate glassware, thereby reducing weight loss and premature breakage.
- Safe and effective cleaner for both glass and plastic labware
- Formulated for use in automatic washers
- Soluble in water at any concentration
- Drains from glassware in seconds
- Contains powerful sequestering reagents
- Ideal for cell culture lab where any trace of contamination can interfere with the growth of cell cultures
- Stable for long-term storage
Specifications
Specifications
| Form | Liquid |
| Appearance | Dark Yellow |
| Content And Storage | Store at Room Temperature |
| Volume (English) | 55 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.