Learn More
Cayman Chemical 8 9-EE-14 Z-E 50ug
SDP
Supplier:  Cayman Chemical 1811250 UG
Encompass_Preferred
An antagonist of NR2C- and NR2D-subunit containing NMDA receptors selective for NR2C- and NR2D-subunit containing NMDA receptors over receptors containing NR2A and NR2B subunits as well as over the AMPA receptor GluR1 (IC50s 229 300 and 300 UM respectively) requires glutamate but not glycine binding for its activity
Catalog No. 50-296-5176
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.