Learn More
8-Aminoquinoline, 98+%
Description
8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.
Solubility
Soluble in water.
Notes
Air Sensitive. Store away from air under dry inert gas. Incompatible with air and oxidizing agents.
Specifications
Specifications
| Melting Point | 63°C to 67°C |
| Boiling Point | 174°C (26 mmHg) |
| Quantity | 5 g |
| Beilstein | 114474 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 144.18 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 8-Aminoquinoline |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335-H341 |
| Hazard Statement | GHS H Statement H341-H315-H319-H335 Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 209-427-9 |
| RTECSNumber | VA9627000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.