Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Anilino-1-naphthalenesulfonic acid, 98%

Catalog No. AC401210250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC401210250 25 g
AC401210050 5 g
AC401211000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC401210250 Supplier Thermo Scientific Chemicals Supplier No. 401210250
Add to Cart

CAS: 82-76-8 | C16H13NO3S | 299.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-76-8
Molecular Formula C16H13NO3S
Molecular Weight (g/mol) 299.34
InChI Key FWEOQOXTVHGIFQ-UHFFFAOYSA-N
Synonym 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid
PubChem CID 1369
ChEBI CHEBI:39708
IUPAC Name 8-anilinonaphthalene-1-sulfonic acid
SMILES C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 183.0°C to 183.5°C
Color Black to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Merck Index 15,654
Formula Weight 299.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 8-Anilino-1-naphthalenesulfonic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-438-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.