Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 8-Azaguanine, Purine Analogue, Antineoplastic, >99% Purity, 50 mg

Catalog No. 502705074
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5074 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5074 Supplier Abcam Supplier No. AB14579850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purine analogue with antineoplastic activity that acts as an antimetabolite to inhibit cellular growth.

  • Molecular weight: 152.11 g/mol
  • Purity: >99%
  • Form: Solid
  • Description: Purine analogue with potential antineoplastic activity and antimetabolite properties. Competes with guanine and incorporates into ribonucleic acids, interfering with biosynthetic pathways to inhibit cellular growth.
  • Storage: Store at -20°C.
  • Shipping: Ambient - Can Ship with Ice
  • Composition: 99.96% 8-Azaguanine

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Guanine
Substrate Properties Chemical Substrate
Content And Storage Store at -20°C.
Buffer 99.96% 8-Azaguanine
Form Solid
Shipping Condition Ambient
Formulation 99.96% 8-Azaguanine
Quantity 50 mg
For Use With (Application) Antineoplastic research, antimetabolite studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.