Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromo-1-octanol 90.0+%, TCI America™
SDP

Catalog No. B172925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B172925G 25 g
B17295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B172925G Supplier TCI America Supplier No. B172925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50816-19-8
Molecular Formula C8H17BrO
Molecular Weight (g/mol) 209.127
MDL Number MFCD00010388
InChI Key GMXIEASXPUEOTG-UHFFFAOYSA-N
Synonym 8-bromo-1-octanol, 1-octanol, 8-bromo, 8-bromooctanol, octamethylene bromohydrin, 8-bromo octanol, 1-bromooctan-8-ol, 8-bromo-1-actanol, 8-bromooctane-1-ol, 1-bromo-octan-8-ol, 8-bromo-octan-1-ol
PubChem CID 162607
IUPAC Name 8-bromooctan-1-ol
SMILES C(CCCCBr)CCCO

Specifications

Color Yellow
Quantity 25 g
Formula Weight 209.13
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Bromo-1-octanol
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.