Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromo-2'-deoxyguanosine 98.0+%, TCI America™
SDP

Catalog No. B4001200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4001200MG 200 mg
B40011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4001200MG Supplier TCI America Supplier No. B4001200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13389-03-2
Molecular Formula C10H12BrN5O4
Molecular Weight (g/mol) 346.14
MDL Number MFCD01630971
InChI Key MKDXZFVCXWXGBQ-QXZWWJJYNA-N
PubChem CID 10947964
ChEBI CHEBI:61072
IUPAC Name 2-amino-8-bromo-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
SMILES NC1=NC2=C(N=C(Br)N2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1

Specifications

Melting Point 210°C
Color White
Quantity 200 mg
Synonym 8-bromo-2'-deoxyguanosine, 8-bromo-2'-dg, 2-amino-8-bromo-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 8-br-2'-dg, guanosine, 8-bromo-2'-deoxy, 8-bromodeoxyguanosine, epitope id:151524, 8-bromo-2?-deoxyguanosine, guanosine,8-bromo-2'-deoxy, 8-bromo-2-deoxyguanosine
Formula Weight 346.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromo-2′-deoxyguanosine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.