Learn More
Apexbio Technology LLC Piribedil 3605-01-4 100mg
SDP
Supplier:  Apexbio Technology LLC B8904100
Encompass_Preferred
Piribedil (CAS 3605-01-4) is a dopamine receptor agonist with affinity primarily for dopamine D2/D3 receptor subtypes mediating dopaminergic neurotransmission By stimulating dopamine receptors this small molecule modulates neuronal signaling pathways relevant to motor control and cognition Piribedil has been studied extensively in preclinical and clinical investigations for its potential utility in treating Parkinson s disease given dopamine signaling deficits characteristic of the disorder It thus serves as a research tool in dopaminergic pharmacology and neurological disorders involving impaired dopamine-mediated neurotransmission
Catalog No. 50-000-63336
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.