Learn More
Medchemexpress LLC 8-bromoadenosine 5'-monophosphate | 23567-96-6 | MFCD00056945 | 98.8% | 426.12 g/mol | C10H13BrN5O7P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1342665MG
Encompass_Preferred
8-bromoadenosine 5'-monophosphate (8-Bromo-AMP; CAS 23567-96-6) is a brominated adenosine monophosphate used as a research reagent and a membrane-permeable cAMP analogue. It is supplied as a solid and is used in biochemical and pharmacological studies to modulate cyclic nucleotide-dependent signaling and to investigate cardiac recovery after ischemia-reperfusion.
  • Membrane-permeable cAMP analogue for signaling studies.
  • High purity, suitable for research applications.
  • Stable brominated adenosine monophosphate analogue.
  • Available in small research quantities such as 5 mg.
  • Useful for studying adenine nucleotide metabolism and cardiac recovery.
Catalog No. 50-002-22847