Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromoadenosine, 98%

Catalog No. AC155780010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155780010 1 g Glass bottle
AC155780050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155780010 Supplier Thermo Scientific Chemicals Supplier No. 155780010
Add to Cart

CAS: 2946-39-6 | C10H12BrN5O4 | 346.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2946-39-6
Molecular Formula C10H12BrN5O4
Molecular Weight (g/mol) 346.14
MDL Number MFCD00005733
InChI Key VJUPMOPLUQHMLE-SBTUYZBDNA-N
Synonym 8-bromoadenosine, bromoadenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, adenosine, 8-bromo, 6-amino-8-bromopurine riboside, 8-bromo-adenosine, 8-bromoadenine-9-beta-d-ribofuranoside, 8-bromo adenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 4s,2r,3r,5r-2-6-amino-8-bromopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol
PubChem CID 96544
ChEBI CHEBI:2312
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-8-bromopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=C(Br)N([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 210°C to 212°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -54.5
Formula Weight 346.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromoadenosine
Show More Show Less
EINECSNumber 220-959-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.