Learn More
Medchemexpress LLC 8-bromoguanosine | 4016-63-1 | MFCD00037985 | 100.0% | 362.14 g/mol | C10H12BrN5O5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01903310G
Encompass_Preferred
8-Bromoguanosine is a brominated derivative of the nucleoside guanosine used as a research reagent to probe RNA structure and function. Supplied as a solid, it can reduce conformational heterogeneity in RNA and is suitable for biochemical and structural biology studies.
  • Brominated guanosine derivative used in RNA research.
  • Reduces RNA conformational heterogeneity to aid structural studies.
  • Supplied as a solid powder for laboratory use.
  • High purity suitable for analytical and structural assays.
  • Applicable to biochemical, biophysical, and structural biology applications.
Catalog No. 50-003-00046