Learn More
Medchemexpress LLC 8-bromoguanosine | 4016-63-1 | MFCD00037985 | 99.6% | 362.14 g·mol⁻¹ | C10H12BrN5O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0190335G
Encompass_Preferred
8-Bromoguanosine is a brominated derivative of the purine nucleoside guanosine used in biochemical and molecular biology research to probe nucleic acid structure and to modulate RNA conformation. It is supplied as a white to off-white solid and in solution formats for laboratory research, and is accompanied by analytical documentation for lot verification.
  • Brominated guanosine analog for nucleic acid studies.
  • Reduces RNA conformational heterogeneity to probe structure and function.
  • High reported purity suitable for analytical and research applications.
  • Available as solid and as concentrated solution for experimental flexibility.
  • Provided with documentation such as COA, SDS, H-NMR, and RP-HPLC for verification.
Catalog No. 50-003-01332