Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromooctanoic Acid 97.0+%, TCI America™
SDP

Catalog No. B167510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B167510G 10 g
B16755G 5 g
B167525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B167510G Supplier TCI America Supplier No. B167510G

Chemical Identifiers

CAS 17696-11-6
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00004430
InChI Key BKJFDZSBZWHRNH-UHFFFAOYSA-N
Synonym 8-bromocaprylic acid, 8-bromo octanoic acid, octanoic acid, 8-bromo, 8-bromo-n-octanoic acid, omega-bromocaprylic acid, 8-bromo-octanoic acid, octanoicacid, 8-bromo, acmc-1bo24, 8-bromooctanoic acid, ksc180g8h
PubChem CID 548275
IUPAC Name 8-bromooctanoic acid
SMILES C(CCCC(=O)O)CCCBr

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 150°C
Quantity 10 g
UN Number 3261
Formula Weight 223.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 8-Bromooctanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.