Learn More
National Safety Apparel™ Enespro™ ArcGuard™ 8 cal Coverall Arc Flash Kit, No Gloves
SDP
The premium choice for superior comfort, improved safety and above average durability
$856.50 - $925.50
Specifications
Ankle Style Open
Arc Flash Rating 8 cal/cm²
Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Closure Type Zipper
Color Navy
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-833
SDP
View Documents
National Safety Apparel™
KIT2CV08NGSM
Small
Each for $856.50
Only null left
Add to Cart
17-001-834
SDP
View Documents
National Safety Apparel™
KIT2CV08NGMD
Medium
Each for $856.50
Only null left
Add to Cart
17-001-835
SDP
View Documents
National Safety Apparel™
KIT2CV08NGLG
Large
Each for $856.50
Only null left
Add to Cart
17-001-836
SDP
View Documents
National Safety Apparel™
KIT2CV08NGXL
X-Large
Each for $856.50
Only null left
Add to Cart
17-001-837
SDP
View Documents
National Safety Apparel™
KIT2CV08NG2X
2X-Large
Each for $880.00
Only null left
Add to Cart
17-001-838
SDP
View Documents
National Safety Apparel™
KIT2CV08NG3X
3X-Large
Each for $880.00
Only null left
Add to Cart
17-001-839
SDP
View Documents
National Safety Apparel™
KIT2CV08NG4X
4X-Large
Each for $901.00
Only null left
Add to Cart
17-001-840
SDP
View Documents
National Safety Apparel™
KIT2CV08NG5X
5X-Large
Each for $925.50
Only null left
Add to Cart
Description
Description
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper
Specifications
Specifications
Open
CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Navy
Arc Flash Kit
None
Unisex
8 cal/cm²
Zipper
Cotton, Nylon
Stitched
Utilities, Manufacturing, Electrical Contractors
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.