Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Chloro-1-n-octanol 97.0+%, TCI America™
SDP

Catalog No. C244825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C244825ML 25 mL
C24485ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C244825ML Supplier TCI America Supplier No. C244825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23144-52-7
Molecular Formula C8H17ClO
Molecular Weight (g/mol) 164.673
MDL Number MFCD00040005
InChI Key YDFAJMDFCCJZSI-UHFFFAOYSA-N
Synonym 8-chloro-1-octanol, 1-octanol, 8-chloro, 8-chlorooctanol, 8-chloro-octan-1-ol, 8-chloro-1-n-octanol, 8-chlor-1-octanol, 8-chlor-octan-1-ol, acmc-1cr4x
PubChem CID 90012
IUPAC Name 8-chlorooctan-1-ol
SMILES C(CCCCCl)CCCO

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
Formula Weight 164.67
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Chloro-1-n-octanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.