Learn More
Abcam 8-CPT-cAMP, Cell Permeable, >99% Purity, Solid, 100 mg
Description
- Molecular weight: 493.8 Da
- Purity: >99%
- Form: Solid
- Key properties: Membrane permeable, lipophilic cAMP analog
- Activity: Activator of Epac, cAMP, and cGMP-dependent protein kinases (PKA/PKG)
- Selectivity: Site B of cAMP-dependent PKA type II
- Inhibition: Inhibits cyclic GMP-specific phosphodiesterase (PDE Va), PDE III, and PDE IV (IC50 values of 0.9, 24, and 25 μM, respectively)
- In vivo effects: Increases heart rate; causes fall in arterial blood pressure and vascular resistance
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% 8-(4-Chlorophenylthio)adenosine 3',5'-cyclic monophosphate |
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Target Molecule | Epac, PKA, PKG, Phosphodiesterases (PDE Va, PDE III, PDE IV) |
| Quantity | 100 mg |
| For Use With (Application) | Activator of Epac, cAMP and cGMP-dependent protein kinases (PKA/PKG), Inhibitor of cyclic GMP-specific phosphodiesterase (PDE Va), PDE III and PDE IV |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.