Learn More
Dual Manufacturing Company Inc Deep Stainless Stell Mesh Sieve | Mesh #30
Supplier:  Dual Manufacturing Company Inc US830FS
Encompass_Preferred
Stainless steel mesh sieve with a 8in diameter and a number 30 mesh. The mess openings are 600 micron.
Catalog No. NC1820797
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.