Learn More
Market Lab Standard Sharps Containers, 8 Gallon, 15.5""""W x 11""""D x 17.75""""H
Supplier:  Market Lab 0033
Encompass_Preferred
Monitor fill-level through clear lids. Horizontal openings allow for safe disposal. Autoclavable and can be incinerated.
Catalog No. NC9591843
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.