Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxy-5-nitroquinoline, 96%, Thermo Scientific™

Catalog No. AAB2446214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2446214 25 g
AAB2446222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2446214 Supplier Thermo Scientific Chemicals Supplier No. B2446214

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4008-48-4
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.158
MDL Number MFCD00006791
InChI Key RJIWZDNTCBHXAL-UHFFFAOYSA-N
Synonym nitroxoline, 5-nitroquinolin-8-ol, 8-hydroxy-5-nitroquinoline, 5-nitro-8-hydroxyquinoline, 5-nitro-8-quinolinol, nitroxolin, noxibiol, 5-nitrox, 5-nitroxine, 5-nitroks
PubChem CID 19910
ChEBI CHEBI:67121
SMILES C1=CC2=C(C=CC(=C2N=C1)O)[N+](=O)[O-]

Specifications

Melting Point 177°C to 182°C
Odor Odorless
Quantity 25 g
Merck Index 14,6655
Formula Weight 190.16
Percent Purity 96%
Chemical Name or Material 8-Hydroxy-5-nitroquinoline
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 223-662-4
RTECSNumber VC8225000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.