Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 8-Hydroxy-DPAT Hydrobromide, 5-HT1a Receptor Agonist, Solid, 50 mg

Catalog No. 502707463
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7463 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7463 Supplier Abcam Supplier No. AB12058250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective 5-HT1a receptor agonist with moderate affinity for 5-HT7 receptors and ability to antagonize 5-HT reuptake.

  • Molecular Weight: 328.29 Da
  • Purity: >95%
  • Form: Solid
  • Activity: Selective 5-HT1a receptor agonist; moderate affinity for 5-HT7 receptors (pKi = 6.6); able to antagonize 5-HT reuptake
  • Chemical Composition: 100% 7-(Dipropylamino)-5,6,7,8-tetrahydro-1-naphthalenol hydrobromide (1:1)
  • CAS Number: 76135-31-4
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at 4°C.
Form Solid
Target Molecule 5-HT1a Receptor
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Formulation 100% 7-(Dipropylamino)-5,6,7,8-tetrahydro-1-naphthalenol hydrobromide (1:1)
Quantity 50 mg
For Use With (Application) 5-HT1a receptor agonism
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.