Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinoline sulfate monohydrate, 98%

Catalog No. p-147490
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148451000 100 g Glass bottle
AC148455000 500 g Glass bottle
AC148450025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC148451000 Supplier Thermo Scientific Chemicals Supplier No. 148451000
Only null left
Add to Cart

CAS: 207386-91-2 | 0·5 H2O | 203.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207386-91-2
Molecular Formula 0·5 H2O
Molecular Weight (g/mol) 203.2
MDL Number MFCD00149610
InChI Key BNCXJZDIJIVJJO-UHFFFAOYSA-N
Synonym 8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 182.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Merck Index 14, 4843
Solubility Information Solubility in water: soluble in water. Other solubilities: slightly soluble in alcohol, insoluble in ether
Formula Weight 203.2
Percent Purity 98%
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 8-Hydroxyquinoline sulfate monohydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.