Learn More
Grainger Threading Tool 3/16 Carbide
Supplier:  Grainger 16T276
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and metric threads. The overall length is 3 inches and it has an American National Standard Stub Acme thread. This durable tool is made in the USA and well-suited for production threading applications.
Catalog No. 50-276-6241
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.