Learn More
Description
- Produced in vivo by both non-cycloxygenase and cyclooxygenase dependant mechanisms from arachidonic acid3-5
- Shown to be a potent vasoconstrictor, potential mediator of hepatorenal syndrome and atherosclerosis
- Mutagen in 3T3 cells and in vascular smooth muscle cells
- Able to modify the fluidity and integrity of membranes
- Short incubation time
- All components stable at 4°C until the kit’s expiration date
Specifications
Specifications
| Assay Range | Intra-Assay: 7.10%, Inter-Assay: 8.30% |
| Calibration Range | 6.1 to 100000pg/mL |
| Description | 8-iso-PGF2-alpha ELISA Kit is a competitive immunoassay for the quantitative determination of free 8-iso-Prostaglandin F2-alpha in biological fluids. |
| Detection Method | Colorimetric |
| Product Type | 8-iso-PGF2-alpha ELISA Kit |
| Sample Type | Cell Culture Supernatant, Tissue, Urine |
| For Use With (Application) | Quantification |
| Regulatory Status | For research use only (RUO) |
| Sample Volume | 100 μL |
| Storage Requirements | Store the kit at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.