Learn More
Enzo Life Sciences 8 iso PGF2 alpha polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADI905016100
Encompass_Preferred
Host: Rabbit. Immunogen: 8-iso-Prostaglandin F2α conjugated to porcine thyroglobulin. Source: Whole rabbit serum. Species reactivity: Species independent. Applications: ELISA. Formulation: Liquid. Contains sodium azide.
Catalog No. 50-200-6901
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.