Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Abnova™ 8-iso-PGF2-α ELISA Kit

Catalog No. 89100187 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-187 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-187 Supplier Abnova™ Supplier No. KA0313
Add to Cart
Add to Cart

8-iso-PGF2-alpha ELISA Kit is a competitive immunoassay for the quantitative determination of 8-iso-PGF2-alpha in biological fluids.

  • Compatible with 8-isoPGF2a in serum, plasma, and tissue samples
  • Shown to be a potent vascoconstrictor, potential mediator of hepatorenal syndrome and etherosclerosis
  • Able to modify the fluidity and integrity of membranes
  • Short incubation time
  • All components stable at 4°C until the kit’s expiration date

Specifications

Assay Range Intra-Assay: 7.36%, Inter-Assay: 7.20%
Calibration Range 160 to 100000pg/mL
Description 8-iso-PGF2-alpha ELISA Kit is a competitive immunoassay for the quantitative determination of 8-iso-PGF2-alpha in biological fluids.
Detection Method Colorimetric
Product Type 8-iso-PGF2-alpha ELISA Kit
Sample Type Plasma, Serum, Tissue
For Use With (Application) Quantification
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 50 μL
Storage Requirements Store the kit at 4°C.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.