Learn More
AdipoGen 8-Prenylnaringenin (5 mg)
SDP
Supplier:  AdipoGen AGCN20525M005
Encompass_Preferred
Chemical. CAS: 53846-50-7. Formula: C20H20O5. MW: 340.4. Prenylflavonoid non-steroidal phytoestrogen that mimicks and/or modulates endogenous estrogens via estrogen receptor binding. Inhibits both isoforms of the human estrogen receptor (ER; IC50s = 57nM and 68nM for ERalpha and ERbeta, respectively). Acts as a full agonist of ERalpha. Cancer chemopreventive agent. Shown to decrease ROS and increase the oxidative phosphorylation system (OXPHOS). Anticancer agent with antiangiogenic and antiproliferative properties. Shown to inhibit cell cycle progression, induce apoptosis in MCF-7 cells and induce autophagy in prostate cancer cell lines. Potent aromatase inhibitor (IC50=65nM) and cellular histone deacetylases (HDACs) inhibitor. Anti-diabetic agent. Anti-osteoporetic agent. Anti-inflammatory and vascularprotective compound. Potential lead structure for potential new therapeutic agents.
Catalog No. 50-205-3698
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.