Learn More
Mascon Medical Mascon Medical™ 8-Strip PCR Tube With Cap, 0.2ml, 125EA Tubes/Bag, 125 EA Caps/Bag, 1 Bag Tubes & 1 Bag Caps/Box, 10 Boxes/Case
SDP
Supplier:  Mascon Medical FSPCRTUBE0.2STRIP
Encompass_Preferred
Mascon Medical™ 8-Strip PCR Tubes are made of high-quality, medical-grade polypropylene suitable for standard PCR heating modules with a reliable sealing performance. This is imperative to reduce liquid loss caused by evaporation while ensuring experimental accuracy. The 8-Strip PCR Tubes are widely used in genetics, biochemistry, immunity, medicine, and other fields, not only for basic research such as gene separation and cloning but also for disease diagnosis or any place using DNA and RNA.Features:• Negative Pressure Detection: -60kpa for 10 minutes• Tolerance Temperature Range: -40°C to 100°C• Centrifugal Force Test: • DNase/RNase free• Non-Pyrogenic• Meets ISO13485 quality management standardSpecifications:• Volume: 0.2mL• Qty. / Case: 125 tubes & 125 caps• Shape: Flat Cap• Color: Transparent• Material: Polypropylene• Size: 77.4*7.6*21.8mm• Packaging Format: 125EA Tubes/Bag, 125 EA Caps/Bag, 1 Bag Tubes & 1 Bag Caps/Box, 10 Boxes/Case
Catalog No. NC2458279
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.