Learn More
Andwin Scientific 8-STRIPS PCR TUBES 125/BX
SDP
Supplier:  Andwin Scientific T30141
Encompass_Preferred
The 8-strip Ultraflux is compatible with most thermal cyclers. Conveniently, there are equal number of tubes and caps in a single pack. The product is specially designed with an ultra thin wall that allows great and quick heat transfer and higher yield. The tubes are pre-numbered, which then allows the user to easily identify the sample. With its crystal-clear design, the content of each well can be easily viewed. It is sold in a box of 125.
Catalog No. NC9923114
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.