Learn More
Grainger 8 Test Sieve Shaker 110 VAC 60 Hz
Supplier:  Grainger 16Y908
Encompass_Preferred
The Advantech Sieve Shaker Conversion Kit converts an Advantech Sieve Shaker to accommodate 8" diameter sieves. It includes an 8" sieve clamp plate to hold 8" sieves and a lid with collar to contain the sample. This conversion kit allows you to use both 8" and 12" sieves on the same shaker, expanding its versatility. The kit is made from stainless steel for durability. It is designed for Advantech sieve shakers models RX-24, RX-29, and RX-29-1. This high quality conversion kit enables you to get more functionality from your Advantech sieve shaker.
Catalog No. 50-276-8058
Only null left
Add to Cart
Request a Quote