Learn More
World Precision Instrument Tilting Base, 5.1"x 0.25"x 2.3"
SDP
Catalog No. 50822967
Encompass
  • Adjustable to 80 degrees
  • 5.1"x 0.25"x 2.3" (60 x 130 x 6mm)
  • 5464 5-lb weight for Tilting Base sold separately
  • Thumbscrew included for metric anti-vibration platforms with M6 holes
Catalog No. 50-822-967 Supplier World Precision Instrument Supplier No. M3
  • Adjustable to 80 degrees
  • 5.1"x 0.25"x 2.3" (60 x 130 x 6mm)
  • 5464 5-lb weight for Tilting Base sold separately
  • Thumbscrew included for metric anti-vibration platforms with M6 holes