Learn More
ELKO FILTERING CO 80 MICRON MESH 1-4 YRDS ELKO
SDP
Supplier:  ELKO FILTERING CO 0380/41
Encompass
Nylon Monofilament Precision Woven Mesh - 80 Micron Opening, Open Area 41%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-80/41
Catalog No. NC0074276
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.