Learn More
Moldex™ 8000 Series Half-Mask P100 Pre-Assembled Respirators
SDP
Catalog No. 19003345A Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-003-345A Small
19-003-345C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-003-345A Supplier Moldex™ Supplier No. 8941
This item is not returnable. View return policy
Pre-assembled 8940 series respirators provide an efficiency level of 99.97% or greater against both oil and non-oil based particulate aerosols. Kit includes 8000 series facepiece, P100 filter disks and filter disk holders.
  • 8000 Series half-mask respirators pre-assembled with P100 particulate filters
  • Soft, durable facepiece and lightweight design with extra-wide sealing
  • Exhalation valve positioned for better communication
  • Cartridges have a snap-in mount for easy replacement and no misthreading
  • Living hinge allows head harness to be folded comfortably around neck when not in use
  • Low profile, compact design fits well under welding helmets and with safety glasses
  • Maintenance made simple with very few replacement parts
  • Fiberglass-free filters, no plastic casing to throw away
1 Includes two 19-003-346 N95 particulate prefilters, two 18-110 retainers, and two 18-102 organic vapor cartridges.
Specifications
Certifications/Compliance ANSI ISEA 110-2003, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR Part 84, TC-23C-1223
Color Gray
Filter Type P100
Material Thermoplastic Elastomer
Product Type Pre-Assembled Half-Mask Respirator
Size Small
Includes 8000 Facepiece. 8940 P100 Particulate Filter Disks and 8900 Filter Disk Holders
Model 8941