Description
- 8000 Series half-mask respirators pre-assembled with P100 particulate filters
- Soft, durable facepiece and lightweight design with extra-wide sealing
- Exhalation valve positioned for better communication
- Cartridges have a snap-in mount for easy replacement and no misthreading
- Living hinge allows head harness to be folded comfortably around neck when not in use
- Low profile, compact design fits well under welding helmets and with safety glasses
- Maintenance made simple with very few replacement parts
- Fiberglass-free filters, no plastic casing to throw away
1 Includes two 19-003-346 N95 particulate prefilters, two 18-110 retainers, and two 18-102 organic vapor cartridges.
Specifications
Specifications
| Certifications/Compliance | ANSI ISEA 110-2003, Section 7.11.1 |
| Certifications (NIOSH) | NIOSH 42 CFR Part 84, TC-23C-1223 |
| Color | Gray |
| Filter Type | P100 |
| Material | Thermoplastic Elastomer |
| Product Type | Pre-Assembled Half-Mask Respirator |
| Size | Large |
| Includes | 8000 Facepiece. 8940 P100 Particulate Filter Disks and 8900 Filter Disk Holders |
| Model | 8943 |