Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 80nm Colloidal Gold Nanoparticles 1OD
SDP

Catalog No. 38010010 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mL
100 mL

Ultra high quality with narrow size distribution and uniform shape

80 nm colloidal gold is an aqueous suspension of spherical metallic nanoparticles manufactured to the highest standard to ensure:
  • Uniform spherical shape
  • Narrow size distribution: 80 nm ± 8nm, <10% CV
  • Ultra high quality
  • Batch-to-batch consistency
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development

Specifications

Quantity 10 mL
Content And Storage 4°C Do not freeze.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.