Learn More
Greiner Bio-One 81- Position Polypropylene Cryo Rack with Datamatrix scanning
Catalog No. p-7075474 Shop All Greiner Bio-One Products
Change view
Click to view available options
Includes:
High-profile Lid
Low-profile Lid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
07-000-938 Low-profile Lid
07-000-937 High-profile Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-938 Supplier Greiner Bio-One Supplier No. 802576
Add to Cart
Add to Cart
Suitable for ultra-low temperature storage of cryogenic vials.
  • Scanning windows underneath each tube
  • Allows for reading of all tube IDs in one scan
  • Rotation stoppers for usage with automated de-/recapping devices
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 4 mL
Description Non sterile
Design Datamatrix Scan
Color Black
Material Polycarbonate
Type Cryo Rack
Includes Low-profile Lid
Quantity 1
Sterility Non-sterile
Dimensions (L x W) 133 x133 mm
Length (Metric) 133 mm
Width (Metric) 133 mm