Learn More
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid
GREENER_CHOICE
Catalog No. 2140224
Change view
Click to view available options
Color:
Fluorescent Green
Fluorescent Orange
Fluorescent Pink
Natural
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-24 Fluorescent Orange
21-402-22 Natural
21-402-23 Fluorescent Green
21-402-25 Fluorescent Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-402-24 Supplier Thermo Scientific™ Supplier No. 8820
Add to Cart
Add to Cart
Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.
Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.
  • Molded with alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
TRUSTED_SUSTAINABILITY
Specifications
Color Fluorescent Orange
Holds 81 (1.5 mL or 2.0 mL Tubes)
Material Polypropylene
Type 81-Well Cryogenic Storage Racks with Lid