Learn More
Avanti Polar Lipids 810892 DSPE PEG(2000)-N- 1MG
Supplier:  Avanti Polar Lipids 810892C1MG
Encompass
810892 | DSPE PEG(2000)-N-Cy7 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[amino(polyethylene glycol)-2000]-N-(Cyanine 7), >99% 1mg CAS#2315262-01-0
Catalog No. NC2032910
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.