Learn More
Enterprise Technology Solutions 8135005796489 Skilcraft Plastic Sheeting, 12 Ft X 100 Ft, Clear
SDP
Supplier:  Enterprise Technology Solutions NSN5796489
Encompass_Preferred
AbilityOne Polyethylene plastic sheeting for general-purpose packaging, surface protection, construction and agricultural uses. It helps protect surfaces from dust and is highly water-resistant and moderately vapor-resistant. Plastic sheeting is not intended for use as food wrapping. Made to LP-378 standards.
Catalog No. 50-206-2795
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.