Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8260 Volatile Organics Kit, Restek™
SDP

Catalog No. 06729222 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-729-222 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-729-222 Supplier Restek Corporation Supplier No. 30076
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Contains 1mL each of these mixtures.

30042: 502.2 Calibration Mix 1 (gases)

  • Bromomethane (74-83-9)
  • Chloroethane (75-00-3)
  • Chloromethane (74-87-3)
  • Dichlorodifluoromethane (CFC-12) (75-71-8)
  • Trichlorofluoromethane (CFC-11) (75-69-4)
  • Vinyl chloride (75-01-4)

30043: 502.2 Calibration Mix 2

  • Bromodichloromethane (75-27-4)
  • Bromoform (75-25-2)
  • Carbon tetrachloride (56-23-5)
  • Chloroform (67-66-3)
  • 1,1-Dichloroethane (75-34-3)
  • 1,1-Dichloroethene (75-35-4)
  • trans-1,2-Dichloroethene (156-60-5)
  • 1,3-Dichloropropane (142-28-9)
  • 2,2-Dichloropropane (594-20-7)
  • cis-1,3-Dichloropropene (10061-01-5)
  • trans-1,3-Dichloropropene (10061-02-6)
  • Methylene chloride (dichloromethane) (75-09-2)
  • 1,1,1-Trichloroethane (71-55-6)
  • Trichloroethene (79-01-6)

30044: 502.2 Calibration Mix 3

  • Bromochloromethane (74-97-5)
  • Dibromochloromethane (124-48-1)
  • 1,2-Dibromo-3-chloropropane (96-12-8)
  • 1,2-Dibromoethane (EDB) (106-93-4)
  • Dibromomethane (74-95-3)
  • 1,2-Dichloroethane (107-06-2)
  • cis-1,2-Dichloroethene (156-59-2)
  • 1,2-Dichloropropane (78-87-5)
  • 1,1-Dichloropropene (563-58-6)
  • 1,1,1,2-Tetrachloroethane (630-20-6)
  • 1,1,2,2-Tetrachloroethane (79-34-5)
  • Tetrachloroethene (127-18-4)
  • 1,1,2-Trichloroethane (79-00-5)
  • 1,2,3-Trichloropropane (96-18-4)

30045: 502.2 Calibration Mix 4

  • Benzene (71-43-2)
  • tert-Butylbenzene (98-06-6)
  • Chlorobenzene (108-90-7)
  • Isopropylbenzene (cumene) (98-82-8)
  • n-Propylbenzene (103-65-1)
  • Styrene (100-42-5)
  • Toluene (108-88-3)
  • 1,3,5-Trimethylbenzene (108-67-8)
  • m-Xylene (108-38-3)

30046: 502.2 Calibration Mix 5

  • Bromobenzene (108-86-1)
  • n-Butylbenzene (104-51-8)
  • sec-Butylbenzene (135-98-8)
  • 2-Chlorotoluene (95-49-8)
  • 1,3-Dichlorobenzene (541-73-1)
  • Ethylbenzene (100-41-4)
  • 1,2,4-Trichlorobenzene (120-82-1)
  • 1,2,4-Trimethylbenzene (95-63-6)
  • o-Xylene (95-47-6)
  • p-Xylene (106-42-3)

30047: 502.2 Calibration Mix 6

  • 4-Chlorotoluene (106-43-4)
  • 1,2-Dichlorobenzene (95-50-1)
  • 1,4-Dichlorobenzene (106-46-7)
  • Hexachlorobutadiene (87-68-3)
  • 4-Isopropyltoluene (p-Cymene) (99-87-6)
  • Naphthalene (91-20-3)
  • 1,2,3-Trichlorobenzene (87-61-6)

30067: 1-Bromo-4-fluorobenzene (BFB)

  • 1-Bromo-4-fluorobenzene (BFB) (460-00-4)

30073: 8260 Surrogate Mix

  • 1-Bromo-4-fluorobenzene (BFB) (460-00-4)
  • Dibromofluoromethane (1868-53-7)
  • Toluene-d8 (2037-26-5)

30074: 8260 Internal Standard Mix

  • Chlorobenzene-d5 (3114-55-4)
  • 1,4-Dichlorobenzene-d4 (3855-82-1)
  • 1,4-Difluorobenzene (540-36-3)
  • Pentafluorobenzene (363-72-4)

30075: 8240/8260 System Performance Check Mix

  • Bromoform (75-25-2)
  • Chlorobenzene (108-90-7)
  • Chloromethane (methyl chloride) (74-87-3)
  • 1,1-Dichloroethane (75-34-3)
  • 1,1,2,2-Tetrachloroethane (79-34-5)

30005: VOA Matrix Spike Mix

  • Benzene (71-43-2)
  • Chlorobenzene (108-90-7)
  • 1,1-Dichloroethene (75-35-4)
  • Toluene (108-88-3)
  • Trichloroethene (79-01-6)

Specifications

Quantity 1 mL
ShelfLife 6 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.