Learn More
DIGI KEY CORPORATION 838AR - Total Ground Carbon Conductive Coating, 15ml
SDP
Supplier:  DIGI KEY CORPORATION 4731413ND
Encompass
This carbon paint consists of a 1-part, solvent-based acrylic lacquer, pigmented with highly conductive carbon powder. It is smooth, durable, and abrasion resistant. It can be easily applied by brush or spray. It has a quick dry time, with no heat cure necessary. It adheres strongly to most injection-molded plastics, such as ABS, PVC, nylon and polycarbonate. It provides strong corrosion resistance and is suitable for use in marine environments.This conductive paint is excellent for use as grounding paint, electrical contact paint, or carbon shielding paint in low frequency RFI shielding applications. It is perfect for shielding metal detectors and other devices where a metal-filled conductive paint would cause malfunction.
Catalog No. NC2883586