Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibutyl sebacate

Catalog No. 848401L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-840-1L 1 L
84-840-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 84-840-1L Supplier Sigma Aldrich Supplier No. 848401L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dibutyl sebacate has been used as a plasticizer to prepare free ethylcellulose films
  • It can be utilized as a core in liquid-core capsules with cross-linked alginate/polyacrylamide membrane prepared via co-extrusion jet-break-up method for the extraction of the pesticide, atrazine

Specifications

CAS 109-43-3
Density 0.936 g/mL (at 25°C (literature))
Boiling Point 178°C to 179°C (3 mmHg)
Molecular Formula C18H34O4
Linear Formula [-(CH2)4 CO2(CH2)3 CH3]2
Refractive Index n20/D 1.441 (literature); n20/D 1.441
MDL Number MFCD00027218
Quantity 1 L
Synonym Decanedioic acid dibutyl ester; Sebacic acid dibutyl ester
Molecular Weight (g/mol) 314.46
Percent Purity ≥97.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.