Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyvinylpyrrolidone

Catalog No. 856568100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-656-8100G 100 g
85-656-8500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 85-656-8100G Supplier Sigma Aldrich Supplier No. 856568100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PVP has biocompatibility, low toxicity, adhesive characteristics, complexing stability, relatively inert behavior, and is resistant to thermal degradation. It is readily prepared by the polymerization of N-vinyl-2-pyrrolidone or by Reppe synthesis technique.PVP is one of the constituents used in the synthesis and coating of silver nanocubes. It serves as a reducing agent and colloidal stabilizer in the synthesis of palladium nanobars, triangular,nanoplates,twinned nanorods and bipyramids. It is also used as a capping agent in the synthesis of silver nanospheres,gold nanoframes. General applications of PVP are: in pharmaceutical, food, beverage, cosmetic, toiletry and photographic industries.Polyvinylpyrrolidone is a component of Denhardt's Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Specifications

CAS 9003-39-8
Linear Formula (C6H9NO)n
MDL Number MFCD00149016
Quantity 100 g
Synonym PVP; Polyvidone; Povidone
Molecular Weight (g/mol) 55000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.