Learn More
Description
- Anhydrous terpineol has been used as a high viscosity solvent along with ethyl cellulose as a resin to prepare dispersion paste of titanium dioxide (tio2) that is used in dye-sensitized solar cell (dssc) technology
- It can also be used in the preparation of graphene/terpineol dispersion for use in inkjet printing.sure/seal is a trademark of sigma-aldrich co
- Llc
Specifications
Specifications
| Density | 0.934 g/mL (at 20°C (literature)) |
| Boiling Point | 213°C to 218°C (lit.) |
| Molecular Formula | C10H18O |
| Refractive Index | n20/D 1.484 |
| Linear Formula | C10H18O |
| MDL Number | MFCD00166983 |
| Quantity | 1 L |
| Molecular Weight (g/mol) | 154.25 |
| Percent Purity | 60.0-85.0% (GC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.