Learn More
PHCbi VIP ECO Natural Refrigerant -86°C Freezer
Supplier:  PHCbi MDFDU702VHAPA1
Encompass_Preferred
PHCbi brand ultra-low temperature (ULT) VIP ECO freezers are engineered from the inside out to create, maintain, and restore precise temperatures. Our purpose-built ultra-low temperature freezers achieve and maintain temperature uniformity and stability not commonly found with household or commercial cabinets.The MDF-DU702VHA-PA provides reliable temperatures as low as -80°C. The MDF-DU702VHA-PA is part of the VIP ECO Series of ultra-low temperature freezers. PHCbi brand VIP ECO Series ULTs have earned a worldwide reputation for dependable performance in demanding environments where critical materials must be preserved for generations.
Catalog No. NC3254458
Request a Quote
Please call customer service for current pricing. 1-800-766-7000