Learn More
Market Lab Specimen Containers, 86 oz, 25/pk
Supplier:  Market Lab 5160
Encompass_Preferred
Stackable design conserves valuable storage space. Withstands formaldehyde, weak acids, and bases. Resistant to freezing and boiling. Nonsterile.
Catalog No. NC1464669
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.