Learn More
Cayman Chemical 8BromoInosIn5OtrIphosph 600ug
SDP
Supplier:  Cayman Chemical 38662600 UG
Encompass_Preferred
A colorimetric peroxidase substrate enzymatic oxidation produces a radical cation followed by a diimine product that can be quantified by colorimetric detection at 652 and 450 nm respectively as a measure of peroxidase activity has commonly been used as a substrate for HRP in ELISA and immunohistochemical applications
Catalog No. 50-291-8761
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.