Learn More
Thermo Scientific™ 8mm Assembled Amber Autosampler Vial Kits
Catalog No. 03378365
Change view
Click to view available options
Closure Material:
Phenolic
Polypropylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Material
03-378-365 Phenolic
03-375-20B Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-378-365 Supplier Thermo Scientific™ Supplier No. C4013494A
Save time and reduce the risk of contamination using this convenience kit. It includes 100 amber vials pre-assembled with 100 screw thread caps and septa.
Save time during sample preparation and reduce the risk of contamination. Each Thermo Scientific™ 8mm Amber Vial Convenience Kit includes 100 amber glass vials preassembled with 100 screw thread caps and septa. They are packaged in convenient vial trays with clear covers or in economical polybags Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Description Amber Glass; Phenolic Black Cap; Red PTFE/White Silicone septum; in storage tray
Color Amber
Material Glass, PTFE, Silicone
Volume (Metric) 2 mL
Septum Red PTFE/White Silicone
No. per Pack 100
Closure Material Phenolic
Closure Type Phenolic
Closure Size 8-425
Marking Spot No
Show More Show Less
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.